Learn More
Biotage SPEFRT PEFRITS 20U/25ML 100
SDP
Catalog No. 502311361
Encompass
Frit; Biotage; Isolute; For SPE; Polyethylene; 20um; 20mm (0.812 in.) dia.; Column type E, 25mL; 100/pack
Catalog No. 50-231-1361 Supplier Biotage Supplier No. 1201037E
Frit; Biotage; Isolute; For SPE; Polyethylene; 20um; 20mm (0.812 in.) dia.; Column type E, 25mL; 100/pack