Learn More
Biotage SPEFRT PEFRITS 20U/3ML 100
SDP
Catalog No. 502311357
Encompass
Frit; Biotage; Isolute; For SPE; Polyethylene; 20um; 9mm (0.375 in.) dia.; Column type B, 3mL; 100/pack
Catalog No. 50-231-1357 Supplier Biotage Supplier No. 1201033B
Frit; Biotage; Isolute; For SPE; Polyethylene; 20um; 9mm (0.375 in.) dia.; Column type B, 3mL; 100/pack