Learn More
Biotage SPEFRT PEFRITS 20U/70ML 100
SDP
Catalog No. 502311362
Encompass
Frit; Biotage; Isolute; For SPE; Polyethylene; 20um; 27mm (1.062 in.) dia.; Column type F, 70mL; 100/pack
Catalog No. 50-231-1362 Supplier Biotage Supplier No. 1201038F
Frit; Biotage; Isolute; For SPE; Polyethylene; 20um; 27mm (1.062 in.) dia.; Column type F, 70mL; 100/pack