Learn More
Analytical Sales & Services Speical 2 Piece Ferrules for 1/16" OD Tubing
Catalog No. 5000806394
Encompass
Speical 2 Piece Ferrules for 1/16" OD Tubing - to be used with Low Pressure Valves, VXL family
Catalog No. 50-008-06394 Supplier Analytical Sales & Services Supplier No. 7770044
Speical 2 Piece Ferrules for 1/16" OD Tubing - to be used with Low Pressure Valves, VXL family