Learn More
Electron Microscopy Sciences Speirs-Levy Eosinophil Counting Chamber
SDP
Catalog No. 5027820
Encompass_Preferred
Speirs-Levy eosinophil is used for counting eosinphil in blood and may also be used for performing cell counts in spinal fluids or other media.
Catalog No. 50-278-20 Supplier Electron Microscopy Sciences Supplier No. 6351256
Speirs-Levy eosinophil is used for counting eosinphil in blood and may also be used for performing cell counts in spinal fluids or other media.