Learn More
Fine Science Tools Spencer Ligature Scissors, Stainless Steel, 9 cm
SDP
Catalog No. NC9056900
Encompass
Tip Shape: Straight/ Tips: Sharp-Sharp/ Cutting Edge: 20mm/ Alloy / Material: Stainless Steel/ Length: 9cm/ Serrated: No
Catalog No. NC9056900 Supplier Fine Science Tools Supplier No. 1407609
Tip Shape: Straight/ Tips: Sharp-Sharp/ Cutting Edge: 20mm/ Alloy / Material: Stainless Steel/ Length: 9cm/ Serrated: No