Learn More
Med Vet International Miltex Vantage Spencer Scissors, 3.5in.
Catalog No. 502829865
Encompass_Preferred
The Miltex Vantage V99-100 Spencer Stitch Scissors are made of mid-grade stainless steel and have a length of 3.5in. These surgical scissors have a small hook shaped tip on one blade that slides under sutures to slightly lift them before cutting for removal.
Catalog No. 50-282-9865 Supplier Med Vet International Supplier No. MILV99100
The Miltex Vantage V99-100 Spencer Stitch Scissors are made of mid-grade stainless steel and have a length of 3.5in. These surgical scissors have a small hook shaped tip on one blade that slides under sutures to slightly lift them before cutting for removal.