Learn More
Sklar Instruments Spencer (Littauer) Stitch Scissors, Sklar, 4-1/2 in., Blunt/Blunt, Straight, Smooth, Finger Rings, Stainless Steel. EA/1.
SDP
Catalog No. 501437861 Shop All Sklar Instruments Products
Encompass_Preferred
Spencer (Littauer) Stitch Scissors, Sklar, 4-1/2 in., Blunt/Blunt, Straight, Smooth, Finger Rings, Stainless Steel. Sklar Spencer Stitch Scissors are primarily used for suture removal. These scissors have a small hook shaped tip on one blade that slides under sutures to slightly lift them before cutting for removal. This hook also holds the suture so it does not slip off the blade of the scissors prior to cutting. They are most commonly available in lengths of 3-1/2 in. and 4-1/2 in. and therefore typically have smaller, more delicate tips that are used for removing smaller sutures. They should not be used for cutting tissue. This product is straight with smooth, blunt/blunt tips and a length of 4-1/2 inches.
This item is not returnable. View return policy
Catalog No. 50-143-7861 Supplier Sklar Instruments Supplier No. 222845
Spencer (Littauer) Stitch Scissors, Sklar, 4-1/2 in., Blunt/Blunt, Straight, Smooth, Finger Rings, Stainless Steel. Sklar Spencer Stitch Scissors are primarily used for suture removal. These scissors have a small hook shaped tip on one blade that slides under sutures to slightly lift them before cutting for removal. This hook also holds the suture so it does not slip off the blade of the scissors prior to cutting. They are most commonly available in lengths of 3-1/2 in. and 4-1/2 in. and therefore typically have smaller, more delicate tips that are used for removing smaller sutures. They should not be used for cutting tissue. This product is straight with smooth, blunt/blunt tips and a length of 4-1/2 inches.