Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Sperian™ Freedom Disposable Respirators

Catalog No. 189992434
Change view
Click to view available options
Filter Type:
Multi-contaminant (organic vapors, acid gas, ammonia, methylamine, formaldehyde)
Organic Vapor (OV)
Includes:
Filter Pad, Organic Vapor Cartridge, Retainer, Reusable Bag
Filter Pad, Retainer, Reusable Bag
Reusable Bag
Size:
Medium
Large
Suspension Type:
Yolk and cradle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Filter Type Includes Size Suspension Type
18-999-2434 Multi-contaminant (organic vapors, acid gas, ammonia, methylamine, formaldehyde) Reusable Bag Medium Yolk and cradle
17-981-27C Organic Vapor (OV) Filter Pad, Organic Vapor Cartridge, Retainer, Reusable Bag Large Yolk and cradle
18-999-2432 Multi-contaminant (organic vapors, acid gas, ammonia, methylamine, formaldehyde) Filter Pad, Retainer, Reusable Bag Large Yolk and cradle
18-999-2431 Multi-contaminant (organic vapors, acid gas, ammonia, methylamine, formaldehyde) Filter Pad, Retainer, Reusable Bag Medium Yolk and cradle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 18-999-2434 Supplier Honeywell™ Supplier No. 14150090

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ideal for occasional users

  • Available in multiple models for a variety of applications
  • Three sizes ensure a secure, comfortable fit
  • Choose from contaminant-specific models or multi-contaminant, multi-use respirators (with cartridge and filter pre-assembled), designed to meet NIOSH standards for multiple contaminant exposure
  • Dispose when cartridges are expended, replacement parts and decontamination concerns are eliminated

Order Info

See chart below for a complete selection of respirators with pre-attached cartridges and filters. Probed models are also available; contact your Fisher Scientific™ Customer Service Representative for details. (Do not exceed maximum use concentration established by regulatory standards.)

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance NIOSH 42CFR84
Color Code Green
Disposable Single-use
Filter Type Multi-contaminant (organic vapors, acid gas, ammonia, methylamine, formaldehyde)
Material Thermoplastic Elastomer
Product Type Half Mask Respirator
Size Medium
For Use With (Application) Fire protection, medical, pharmaceutical, transportation
Includes Reusable Bag
Material (Faceshield) Thermoplastic Elastomer
Quantity 1 Ea.
Suspension Type Yolk and cradle
Weight (English) 0.75 lb.
Weight (Metric) 0.34 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.