Learn More
Honeywell™ Sperian™ One-Fit™ N95 Particulate Respirators
Catalog No. 19034920
Change view
Click to view available options
Size:
Standard
Quantity:
1 Ea.
Catalog No. Size Quantity
19-034-920 Standard 1 Ea.
1 options
This item is not returnable. View return policy
Catalog No. 19-034-920 Supplier Honeywell™ Supplier No. 14110444
This item is not returnable. View return policy
Universal sized disposable respirators fit most face shapes and sizes.
  • Electrostatic nonwoven polypropylene and polyester materials; lightweight construction allows for longer wear time and less fatigue
  • Durable filtration media ensures filtration efficiency of 95% against non-oil airborne particles
  • Protection against airborne pathogens and M. tuberculosis as well as certain airborne particulates and pandemic diseases such as H5N1 Avian Flu and SARs
W1400 Flat Fold Mask
  • Bonded wire nose clip with open cell foam nose seal
  • Low-profile design offers compatibility with spectacles
NBW95 and NBW95V Form Fitted Masks
  • Strong and rigid outer shell prevents mask from collapse in high humidity or moist environments
  • Molded nose bridge fits comfortably and is compatible with spectacles
TRUSTED_SUSTAINABILITY
Specifications
Aerosol Type N - Not Resistant to Oil
Certifications (NIOSH) NIOSH 42 CFR 84, N95, approval number: TC-84A-4373
Color Code White
Color White
Design Type Cup/Molded
Disposable Single-use
Filter Type N95
Material Polypropylene, Polyester
Product Type General Purpose Particulate Respirator
Size Standard
With Exhalation Valve Yes
Filter Efficiency 95%
Latex Free Yes
Material Headstrap Elastic
Model 14110444
Quantity 1 Ea.
Strap Attachment Type Dual Welded Head Straps
FDA Cleared Surgical Mask No