Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Sperian™ Survivair 2000 Series Half-Mask Respirators

Catalog No. 18245A
Change view
Click to view available options
Size:
Small
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-245A Small
18-245C Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 18-245A Supplier Honeywell™ Supplier No. B250000

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

This item is not returnable. View return policy

Silicone reusable half-mask respirator features a lightweight and innovative yoke design with capability to convert to a CF-SAR.

  • Head harness designed to reduce facepiece slippage
  • Silicone facepiece construction is lightweight and durable
  • Rear split strap for easy donning and better support
  • Compatible with S-Series filters and cartridges
TRUSTED_SUSTAINABILITY

Specifications

Certifications (NIOSH) NIOSH 42 CFR Part 84
Color Blue
Material Silicone
Product Type Half-Mask Respirator
Size Small
For Use With (Equipment) S series filter and cartridge
Model B250000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.