Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Sperian™ Survivair Panther™ / Cougar™ SCBA Regulator Accessory, Spring

Catalog No. 17377818
Change view
Click to view available options
Spring; H.P.; 4500psig; Green;
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
17-377-818 Spring; H.P.; 4500psig; Green;
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-377-818 Supplier Honeywell™ Supplier No. 910035

Specifications

Color Green
For Use With (Equipment) Survivair Panther, Cougar Self Contained Breathing Apparatus Regulator
Model 910035
Pressure 4500 psi
Product Type Spring
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.