Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Sperian™ S-Series Cartridges and Filters

Catalog No. 18266A
Change view
Click to view available options
Cartridge Type:
Acid gas with P100
Ammonia, Chlorine Dioxide, Chlorine, Hydrogen Chloride, Hydrogen Fluoride, Hydrogen Sulfide (escape only), Formaldehyde, Methylamine, organic Vapors, or Sulfur Dioxide, all particulate Aerosols Cartridge
Ammonia, Methylamine Cartridge
Organic Vapor Cartridge
Color:
Black/Magenta
Green/Magenta
Olive/Magenta
For Use With (Application):
For Chlorine/Hydrogen Chloride/Sulfur Dioxide/Formaldehyde/P100
For protection against ammonia, chlorine dioxide, chlorine, hydrogen chloride, hydrogen fluoride, hydrogen sulfide, formaldehyde, methylamine, organic vapors, or sulfur dioxide
For protection against ammonia, methylamine, 99.97% efficient against all non-oil and oil based particulates
For protection against organic vapors
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cartridge Type Color For Use With (Application)
18-266A Ammonia, Methylamine Cartridge Green/Magenta For protection against ammonia, methylamine, 99.97% efficient against all non-oil and oil based particulates
18-267B Ammonia, Chlorine Dioxide, Chlorine, Hydrogen Chloride, Hydrogen Fluoride, Hydrogen Sulfide (escape only), Formaldehyde, Methylamine, organic Vapors, or Sulfur Dioxide, all particulate Aerosols Cartridge Olive/Magenta For protection against ammonia, chlorine dioxide, chlorine, hydrogen chloride, hydrogen fluoride, hydrogen sulfide, formaldehyde, methylamine, organic vapors, or sulfur dioxide
18-263A Organic Vapor Cartridge Black/Magenta For protection against organic vapors
18-264A Acid gas with P100 - For Chlorine/Hydrogen Chloride/Sulfur Dioxide/Formaldehyde/P100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-266A Supplier Honeywell™ Supplier No. 105410

S-Series filters work with both full and half mask respirators that only accept threaded cartridge connections

Survivair S-Series Screw-On Cartridges thread easy for tightening cartridge on seal.

Safety/5, † OSHA regulations require gasproof goggles be worn when half-mask respirators are used for protection against formaldehyde.

TRUSTED_SUSTAINABILITY

Specifications

Cartridge Type Ammonia, Methylamine Cartridge
Certifications/Compliance NIOSH
Connection Type Threaded
Color Green/Magenta
Filter Type P100
Product Type Cartridge/Filter Combination
Filter Efficiency 99.97%
For Use With (Application) For protection against ammonia, methylamine, 99.97% efficient against all non-oil and oil based particulates
For Use With (Equipment) Half mask and full facepiece respirator
Quantity 4/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.