Learn More
Honeywell™ Sperian™ Survivair S-Series Respirator Cartridges, Filters, and Cartridge/Filter Combinations
Catalog No. 18255A
Change view
Click to view available options
Cartridge Type:
Acid Gas Cartridge
Organic Vapor Cartridge
Organic Vapor, Acid Gas Cartridge
Quantity:
6/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cartridge Type Quantity
18-255A Organic Vapor Cartridge 6/Pk.
18-257A Organic Vapor, Acid Gas Cartridge 6/Pk.
18-256A Acid Gas Cartridge 6/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 18-255A Supplier Honeywell™ Supplier No. B100100
This item is not returnable. View return policy
Survivair S-Series Screw-On Cartridges thread easy for tightening cartridge on seal.
(Do not exceed maximum use concentration established by regulatory standards.)
Note: OSHA regulations require gasproof goggles be worn when half-mask respirators are used for protection against formaldehyde.
TRUSTED_SUSTAINABILITY
Specifications
Cartridge Type Organic Vapor Cartridge
Product Type Cartridge
Quantity 6/Pk.