Learn More
Honeywell™ Sperian™ Survivair T-Series Cartridges
Catalog No. 176351
Change view
Click to view available options
Cartridge Type:
Mercury Vapor
Organic Vapor
Organic Vapor, Acid Gas
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cartridge Type
17-635-1 Organic Vapor
17-635-3 Organic Vapor, Acid Gas
18-063 Mercury Vapor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 17-635-1 Supplier Honeywell™ Supplier No. T100100
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
This item is not returnable. View return policy
Survivair T-Series twist-lock cartridges provide an audible click indicating a positive lock on connector.
  • Cartridges designed to protect against select organic vapors and acid gases
  • Selection is dependent on type of contaminant, level of exposure, and length of exposure
  • Twist-Lock Cartridges provide an audible click indicating a positive lock on connector
  • 6 cartridges per pack
TRUSTED_SUSTAINABILITY
Specifications
Cartridge Type Organic Vapor
Certifications (NIOSH) NIOSH 42 CFR 84, TC-23C-741
Product Type Cartridge
For Use With (Equipment) Sperian™ T-Series Facepieces
Quantity 6/Pk.