Learn More
Honeywell™ Sperian™ Survivair T-Series Cartridges
Catalog No. 18063
Change view
Click to view available options
Cartridge Type:
Mercury Vapor
Organic Vapor
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cartridge Type
18-063 Mercury Vapor
17-635-1 Organic Vapor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 18-063 Supplier Honeywell™ Supplier No. T100600
This item is not returnable. View return policy
Survivair T-Series twist-lock cartridges provide an audible click indicating a positive lock on connector.
  • Cartridges designed to protect against select organic vapors and acid gases
  • Selection is dependent on type of contaminant, level of exposure, and length of exposure
  • Twist-Lock Cartridges provide an audible click indicating a positive lock on connector
  • 6 cartridges per pack
TRUSTED_SUSTAINABILITY
Specifications
Cartridge Type Mercury Vapor
Certifications (NIOSH) NIOSH 42 CFR 84, TC-23C-741
Product Type Cartridge
For Use With (Equipment) Sperian™ T-Series Facepieces
Quantity 6/Pk.