Learn More
Honeywell™ Sperian™ ValuAir Plus Respirators
Catalog No. 189992429
Change view
Click to view available options
Size:
Small
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-999-2429 Large
18-999-2427 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 18-999-2429 Supplier Honeywell™ Supplier No. 303000
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
Thermoplastic half-mask respirators are intended for limited life applications without sacrificing comfort or fit.
  • Thermoplastic elastomer facepiece features a soft sealing flange with rolled edges
  • Forward-angled nosepiece for compatibility with protective eyewear
  • Perspiration port drains excess moisture off the face, reducing slippage and irritation
  • Easy donning with three-piece sliding suspension system—no hooks, loops, or extra parts
  • Two-point headband suspension adjustment keeps straps out of the field of vision
Uses Survivair S-Series cartridges and filters.
TRUSTED_SUSTAINABILITY
Specifications
Material Thermoplastic Elastomer
Product Type Half-Mask Respirator
Size Large
For Use With (Application) Paint and pesticides
For Use With (Equipment) T-Series Cartridges and Filters
Model 302000