Learn More
Honeywell Sperian VisitorSpec
Description
Hazards
- Abrasion
- Airborne Particulates
- Biohazard
- Impact
Certifications
ANSI Z87+ - Meets ANSI Z87+ standard; CSA Z94.3 - Tested by Honeywell to meet the requirements of the CSA Z94.3 standard
Specifications
Specifications
| Lens Type | Uni-lens |
| Certifications/Compliance | ANSI Z87+; CSA Z94.3 |
| Frame Color | Black |
| Temple Style | 1 Piece |
| Weight (Metric) | 41.4 g |
| Lens Material | Polycarbonate |
| Frame Type | No frame, uni-lens |