Learn More
NEXPRING HEALTH SPERM WASHING MEDIUM 100ML EA
Catalog No. NC0231898
Encompass
Sperm Washing Medium, 100ML Developed to maintain sperm quality during in vitro washing procedures and sperm preparation outside the incubator. Sperm Washing Medium supports all steps of sperm preparation prior to fertilization, whether it is IUI, IVF, or other ART procedures. It is designed to maintain sperm quality on the tabletop without CO2 incubation. Sperm Washing Medium provides lot-to-lot consistency for sperm processing through its quality formulation, which is stringently tested by mouse embryo assay (MEA). • Formulation: HEPES-buffered HTF with human serum albumin • Does not contain antibiotics • CE Marked Quality Control Testing: • Endotoxin by Limulus Amebocyte Lysate (LAL) • Mouse Embryo Assay • Sterility by the current USP, CFR or Ph. Eur. Sterility Test • pH • Osmolality
Catalog No. NC0231898 Supplier NEXPRING HEALTH Supplier No. 9983100ML
Sperm Washing Medium, 100ML Developed to maintain sperm quality during in vitro washing procedures and sperm preparation outside the incubator. Sperm Washing Medium supports all steps of sperm preparation prior to fertilization, whether it is IUI, IVF, or other ART procedures. It is designed to maintain sperm quality on the tabletop without CO2 incubation. Sperm Washing Medium provides lot-to-lot consistency for sperm processing through its quality formulation, which is stringently tested by mouse embryo assay (MEA). • Formulation: HEPES-buffered HTF with human serum albumin • Does not contain antibiotics • CE Marked Quality Control Testing: • Endotoxin by Limulus Amebocyte Lysate (LAL) • Mouse Embryo Assay • Sterility by the current USP, CFR or Ph. Eur. Sterility Test • pH • Osmolality