Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

InVitroCare™ SpermCare™ One Step Sperm Preparation System

Catalog No. 23000917
Encompass
Encompass_Preferred
Change view
Click to view available options
For Use With (Application):
Intrauterine insemination
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
23-000-917 Intrauterine insemination
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-000-917 Supplier InVitroCare™ Supplier No. 2224
May include imposed supplier surcharges.
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Pre-measured gradient in 15mL centrifuge tubes, pre-mesured InVitroCare sperm wash medium in glass vials

Ready-to-use, pre-measured gradient system for sperm preparation for use in INTRAUTERINE INSEMINATION (IUI).

  • Sperm isolated by layering 1 – 2mL semen on top of SpermCare™ gradient and centrifuging for 30 min. at 500xg
  • w/Gradient consisting of silane coated colloidal silica particles suspended in HEPES-buffered HTF
  • Pre-measured containers reduce chance of contamination and help eliminate product waste
  • Contains glucose as usable energy susbtrate to maintain normal cell metabolism, sperm function
  • Aseptically packaged and terminally sterilized
  • Contains product for five determinations
  • Shelf life: 18 months

Specifications

Content And Storage 2 to 8°C
Product Type Sperm Preparation System
Sterility Sterile
For Use With (Application) Intrauterine insemination
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.