Learn More
Cayman Chemical Spermidine-d6, Purity: ≥99% deuterated forms (d1-d6), CAS: 2514812-10-1, 1 mg
SDP
Catalog No. 502912330
Encompass_Preferred
An internal standard for the quantification of spermidine by GC- or LC-MS
Catalog No. 50-291-2330 Supplier Cayman Chemical Supplier No. 346971 MG
An internal standard for the quantification of spermidine by GC- or LC-MS