Learn More
Medchemexpress LLC Spermidine | 124-20-9 | 99.0% | 25 G
SDP
Catalog No. 5000413874
Encompass_Preferred
Spermidine maintains cell membrane stability, increases antioxidant enzyme activities, improving photosystem II (PSII), and relevant gene expression. It significantly decreases H2O2 and O2.- contents and plays a crucial role against abiotic stresses, such as salt, drought, heat, and salinity-alkalinity stresses in plants like tomato, cucumber, and rice.
  • Maintains cell membrane stability
  • Increases antioxidant enzyme activities
  • Improves photosystem II (PSII)
  • Decreases H2O2 and O2.- contents
  • Plays a crucial role against abiotic stresses
Catalog No. 50-004-13874 Supplier Medchemexpress LLC Supplier No. 25GHYB1776
Spermidine maintains cell membrane stability, increases antioxidant enzyme activities, improving photosystem II (PSII), and relevant gene expression. It significantly decreases H2O2 and O2.- contents and plays a crucial role against abiotic stresses, such as salt, drought, heat, and salinity-alkalinity stresses in plants like tomato, cucumber, and rice.
  • Maintains cell membrane stability
  • Increases antioxidant enzyme activities
  • Improves photosystem II (PSII)
  • Decreases H2O2 and O2.- contents
  • Plays a crucial role against abiotic stresses