Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spermidine, 99%

Catalog No. p-3737385
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC132740010 1 g Glass bottle
AC132740050 5 g Glass bottle
AC132740250 25 g Glass bottle
AC132741000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC132740010 Supplier Thermo Scientific Chemicals Supplier No. 132740010
Only null left
Add to Cart

CAS: 124-20-9 | C7H19N3 | 145.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-20-9
Molecular Formula C7H19N3
Molecular Weight (g/mol) 145.25
MDL Number MFCD00008229
InChI Key ATHGHQPFGPMSJY-UHFFFAOYSA-N
Synonym spermidine, 1,5,10-triazadecane, 4-azaoctamethylenediamine, spermidin, 4-azaoctane-1,8-diamine, n1-3-aminopropyl butane-1,4-diamine, 1,4-butanediamine, n-3-aminopropyl, n-3-aminopropyl butane-1,4-diamine, n-3-aminopropyl-1,4-butane-diamine, 1,4-diaminobutane, n-3-aminopropyl
PubChem CID 1102
ChEBI CHEBI:16610
SMILES NCCCCNCCCN

Specifications

Melting Point 22.0°C to 25.0°C
Color Colorless
Density 0.9250g/mL
Boiling Point 128.0°C to 130.0°C (14.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4785 to 1.4805
Quantity 1 g
Beilstein 04,II,704
Merck Index 15,8869
Specific Gravity 0.925
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol and ether
IUPAC Name N'-(3-aminopropyl)butane-1,4-diamine
Formula Weight 145.25
Percent Purity 99%
Physical Form Liquid After Melting
Chemical Name or Material Spermidine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-689-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.