Learn More
Medchemexpress LLC Spermidine-d6 | 2514812-10-1 | 96.1% | 1 MG
SDP
Catalog No. 5000413220
Encompass_Preferred
Spermidine-d6 is a deuterium-labeled spermidine that helps maintain cell membrane stability, increases antioxidant enzyme activities, improves photosystem II (PSII), and enhances relevant gene expression. It significantly reduces H2O2 and O2.- contents.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-13220 Supplier Medchemexpress LLC Supplier No. HYB1776S1MG
Spermidine-d6 is a deuterium-labeled spermidine that helps maintain cell membrane stability, increases antioxidant enzyme activities, improves photosystem II (PSII), and enhances relevant gene expression. It significantly reduces H2O2 and O2.- contents.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS