Learn More
Medchemexpress LLC Spermidine-d6 | 2514812-10-1 | 96.1% | 151.28 | C7H13D6N3 | 10 MG
SDP
Catalog No. 5000298110
Encompass_Preferred
Spermidine-d6 is a deuterium-labeled analog of spermidine supplied for use as a stable isotope tracer and internal standard in quantitative analytical methods (NMR, GC-MS, LC-MS). It is intended for research use and is provided with high purity and aqueous solubility for analytical workflows.
  • Used as an internal standard for quantitative analysis.
  • Deuterium labeling supports metabolic and tracer studies.
  • High aqueous solubility (≥50 mg/mL) for convenient preparation.
  • Stable under recommended storage conditions for long-term use.
  • Offered in small package sizes suitable for analytical assays.
Catalog No. 50-002-98110 Supplier Medchemexpress LLC Supplier No. HYB1776S10MG
Spermidine-d6 is a deuterium-labeled analog of spermidine supplied for use as a stable isotope tracer and internal standard in quantitative analytical methods (NMR, GC-MS, LC-MS). It is intended for research use and is provided with high purity and aqueous solubility for analytical workflows.
  • Used as an internal standard for quantitative analysis.
  • Deuterium labeling supports metabolic and tracer studies.
  • High aqueous solubility (≥50 mg/mL) for convenient preparation.
  • Stable under recommended storage conditions for long-term use.
  • Offered in small package sizes suitable for analytical assays.