Learn More
Medchemexpress LLC Spermidine-d8 hydrochloride | 1173019-26-5 | 95.0% | 1 MG
SDP
Catalog No. 5000412715
Encompass_Preferred
Spermidine-d8 (hydrochloride) is the deuterium labeled Spermidine trihydrochloride. Spermidine hydrochloride helps maintain cell membrane stability, increases antioxidant enzyme activities, improves photosystem II (PSII), and enhances relevant gene expression. It also significantly decreases H2O2 and O2.- contents. This compound is intended for research use only, and is not sold to patients.
  • Used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-12715 Supplier Medchemexpress LLC Supplier No. HYB1776AS1MG
Spermidine-d8 (hydrochloride) is the deuterium labeled Spermidine trihydrochloride. Spermidine hydrochloride helps maintain cell membrane stability, increases antioxidant enzyme activities, improves photosystem II (PSII), and enhances relevant gene expression. It also significantly decreases H2O2 and O2.- contents. This compound is intended for research use only, and is not sold to patients.
  • Used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS