Learn More
Medchemexpress LLC Spermidine hydrochloride | 334-50-9 | 98.0% | 1 ML
SDP
Catalog No. 5000411521
Encompass_Preferred
Spermidine hydrochloride maintains cell membrane stability, increases antioxidant enzyme activities, and improves photosystem II (PSII) and relevant gene expression. It significantly decreases H2O2 and O2.- contents. It also plays a crucial role against abiotic stresses such as salt, drought, heat, and salinity-alkalinity stresses.
  • Maintains cell membrane stability.
  • Increases antioxidant enzyme activities.
  • Improves photosystem II (PSII) and relevant gene expression.
  • Decreases H2O2 and O2.- contents.
  • Plays a crucial role against abiotic stresses.
Catalog No. 50-004-11521 Supplier Medchemexpress LLC Supplier No. HYB1776A10MM/1ML
Spermidine hydrochloride maintains cell membrane stability, increases antioxidant enzyme activities, and improves photosystem II (PSII) and relevant gene expression. It significantly decreases H2O2 and O2.- contents. It also plays a crucial role against abiotic stresses such as salt, drought, heat, and salinity-alkalinity stresses.
  • Maintains cell membrane stability.
  • Increases antioxidant enzyme activities.
  • Improves photosystem II (PSII) and relevant gene expression.
  • Decreases H2O2 and O2.- contents.
  • Plays a crucial role against abiotic stresses.