Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Spermidine phosphate salt hexahydrate

Catalog No. S03811G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S03811G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S03811G Supplier Sigma Aldrich Supplier No. S03811G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Spermidine phosphate salt hexahydrate

Specifications

CAS 23273-82-7
Molecular Formula C7H19N3 -+ 1.5H3PO4 -+ 3H2O
Linear Formula [NH2(CH2)3 NH(CH2)4 NH2]2 -+ 3H3PO4 -+ 6H2O
MDL Number MFCD04966492
Quantity 10 g
Synonym Dispermidine triphosphate
Molecular Weight (g/mol) 346.28
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.