Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spermidine Trihydrochloride 99% Crystalline MP Biomedicals

Catalog No. ICN10047201 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10047201 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10047201 Supplier MP Biomedicals, Inc Supplier No. 0210047201
Add to Cart
Add to Cart

Chemical Identifiers

CAS 334-50-9
Molecular Formula C7H22Cl3N3
Molecular Weight (g/mol) 254.62
MDL Number MFCD00012918
InChI Key LCNBIHVSOPXFMR-UHFFFAOYSA-N
Synonym spermidine trihydrochloride, n-3-aminopropyl-1,4-butanediamine trihydrochloride, spermidine hydrochloride, n1-3-aminopropyl butane-1,4-diamine trihydrochloride, unii-1o14bed398, 1,4-butanediamine, n-3-aminopropyl-, trihydrochloride, spermidine, trihydrochloride, 1,4-butanediamine, n-3-aminopropyl , hydrochloride, n-3-aminopropyl butane-1,4-diamine trihydrochloride, 4-azoniaoctamethylenediammonium trichloride
PubChem CID 9539
IUPAC Name trihydrogen (4-aminobutyl)(3-aminopropyl)amine trichloride
SMILES [H+].[H+].[H+].[Cl-].[Cl-].[Cl-].NCCCCNCCCN

Specifications

Assay Percent Range 99%
Quantity 1 g
Formula Weight 254.6
Physical Form Crystalline
Chemical Name or Material Spermidine Trihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.