Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spermidine Trihydrochloride, Fisher BioReagents

Catalog No. BP25401 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP25401 1 g Amber Glass
BP25405 5 g Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP25401

Spermidine is an aliphatic polyamine used in various biochemical applications due to its involvement in many metabolic functions within organisms

Forms stable compounds with nucleic acids and may be used for the purification of DNA-binding proteins

Spermidine Trihydrochloride forms stable compounds with nucleic acids. It is used in biochemical research.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 334-50-9
Molecular Formula C7H22Cl3N3
Molecular Weight (g/mol) 254.62
MDL Number MFCD00012918
InChI Key LCNBIHVSOPXFMR-UHFFFAOYSA-N
Synonym spermidine trihydrochloride, n-3-aminopropyl-1,4-butanediamine trihydrochloride, spermidine hydrochloride, n1-3-aminopropyl butane-1,4-diamine trihydrochloride, unii-1o14bed398, 1,4-butanediamine, n-3-aminopropyl-, trihydrochloride, spermidine, trihydrochloride, 1,4-butanediamine, n-3-aminopropyl , hydrochloride, n-3-aminopropyl butane-1,4-diamine trihydrochloride, 4-azoniaoctamethylenediammonium trichloride
PubChem CID 9539
IUPAC Name N'-(3-aminopropyl)butane-1,4-diamine;trihydrochloride
SMILES [H+].[H+].[H+].[Cl-].[Cl-].[Cl-].NCCCCNCCCN

Specifications

Melting Point 248°C
Color White
pH 3 to 6
Assay Percent Range ≥99 %
Packaging Amber Glass
Quantity 1 g
Merck Index 15, 8869
Formula Weight 254.63
Percent Purity 99% min.
Physical Form Solid
Chemical Name or Material Spermidine Trihydrochloride
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Irritating to eyes, respiratory system and skin. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:2
Flammability:1
Instability:1
Recommended Storage Store below 20°C, desiccate.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.