Learn More
Cayman Chemical Spermine NONOate, Purity: ≥98%, CAS: 136587-13-8, 100 mg
SDP
Catalog No. 502896602
Encompass_Preferred
Spermine NONOate is a NO donor. It spontaneously dissociates in a pH-dependent, first-order process with a half-life of 39 minutes and 230 minutes at 37°C and 22-25°C, respectively, (pH 7.4) to liberate 2 moles of NO per mole of parent compound.{875,5437}
Catalog No. 50-289-6602 Supplier Cayman Chemical Supplier No. 82150100 MG
Spermine NONOate is a NO donor. It spontaneously dissociates in a pH-dependent, first-order process with a half-life of 39 minutes and 230 minutes at 37°C and 22-25°C, respectively, (pH 7.4) to liberate 2 moles of NO per mole of parent compound.{875,5437}