Learn More
Medchemexpress LLC HY-B1777 100mg Medchemexpress, Spermine CAS:71-44-3 Purity:>98%
SDP
Catalog No. 501933006
Encompass_Preferred
Medchemexpress, HY-B1777 100mg Spermine CAS:71-44-3 Spermine (NSC 268508) functions directly as a free radical scabenger to protect DNA from free radical attack. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3006 Supplier Medchemexpress LLC Supplier No. HYB1777100MG
Medchemexpress, HY-B1777 100mg Spermine CAS:71-44-3 Spermine (NSC 268508) functions directly as a free radical scabenger to protect DNA from free radical attack. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.