Learn More
Cayman Chemical Spermine, 71-44-3, 1 g
SDP
Supplier:  Cayman Chemical 180411
Encompass_Preferred
An endogenous polyamine required for eukaryotic cell growth and protein synthesis; blocks inward rectifying K+ channels and acts as a mixed NMDA glutamate receptor agonist/antagonist at the polyamine site; scavenges free radicals to prevent DNA damage by ROS
Catalog No. NC1251061