Learn More
Selleck Chemical LLC Spermine 25mg 71-44-3 4,9-Diaza-1,12-dodecanediamine
SDP
Supplier:  Selleck Chemical LLC S394825MG
Encompass_Preferred
Spermine (4,9-Diaza-1,12-dodecanediamine) is a polyamine involved in cellular metabolism found in all eukaryotic cells. It is found in a wide variety of organisms and tissues and is an essential growth factor in some bacteria. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8428