Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Spermine-d20 | 71-44-3 | 98.5% | 222.46 | C10H6D20N4 | 5 MG
SDP

Catalog No. 5000284537
Encompass_Preferred

Spermine-d20 is a deuterium-labeled stable isotope of the endogenous polyamine spermine, supplied as a solid powder for analytical and biochemical research applications, including use as an internal standard for mass spectrometry and tracer studies.

  • Deuterium-labeled analog for accurate mass spectrometry standards.
  • High chemical purity: 98.5%.
  • Molecular weight 222.46 and formula C10H6D20N4 for exact-mass calculations.
  • White to off-white solid form, convenient for handling and storage.
  • Storage recommendations: powder at -20°C (stable for 3 years); in solvent at -80°C for 6 months or -20°C for 1 month.

Catalog No. 50-002-84537 Supplier Medchemexpress LLC Supplier No. HYW6543425MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Spermine-d20 is a deuterium-labeled stable isotope of the endogenous polyamine spermine, supplied as a solid powder for analytical and biochemical research applications, including use as an internal standard for mass spectrometry and tracer studies.

  • Deuterium-labeled analog for accurate mass spectrometry standards.
  • High chemical purity: 98.5%.
  • Molecular weight 222.46 and formula C10H6D20N4 for exact-mass calculations.
  • White to off-white solid form, convenient for handling and storage.
  • Storage recommendations: powder at -20°C (stable for 3 years); in solvent at -80°C for 6 months or -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.