Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Spermine-d20 | 71-44-3 | 98.5% | 222.46 g/mol | C10H6D20N4 | 1 MG
SDP

Catalog No. 5000284808
Encompass_Preferred

Spermine-d20 is a deuterium-labeled isotopologue of spermine used as a stable isotope tracer or internal standard for quantitative NMR, GC-MS, and LC-MS analyses. It provides a mass-shifted analog for accurate quantitation in analytical and tracer studies.

  • Purity 98.47%.
  • Molecular formula C10H6D20N4; molecular weight 222.46 g/mol.
  • Used as a tracer or internal standard for NMR, GC-MS, and LC-MS quantitation.
  • Appearance white to off-white solid.
  • Storage: powder -20°C (3 years); in solution -80°C (6 months), -20°C (1 month).
  • Typical analytical pack size available: 1 mg.

Catalog No. 50-002-84808 Supplier Medchemexpress LLC Supplier No. HYW6543421MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Spermine-d20 is a deuterium-labeled isotopologue of spermine used as a stable isotope tracer or internal standard for quantitative NMR, GC-MS, and LC-MS analyses. It provides a mass-shifted analog for accurate quantitation in analytical and tracer studies.

  • Purity 98.47%.
  • Molecular formula C10H6D20N4; molecular weight 222.46 g/mol.
  • Used as a tracer or internal standard for NMR, GC-MS, and LC-MS quantitation.
  • Appearance white to off-white solid.
  • Storage: powder -20°C (3 years); in solution -80°C (6 months), -20°C (1 month).
  • Typical analytical pack size available: 1 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.