Learn More
Medchemexpress LLC Spermine-d20 | 71-44-3 | 98.5% | 222.46 g/mol | C10H6D20N4 | 25 MG
SDP
Catalog No. 5000284820
Encompass_Preferred
Spermine-d20 is a deuterium-labeled isotopologue of the polyamine spermine supplied as a research-grade stable isotope standard for analytical and biological studies. It is intended for use as an internal standard, tracer, or reference material in mass spectrometry and metabolomics. For research use only.
  • Deuterium-labeled standard for isotope dilution and tracer studies.
  • High purity suitable for analytical applications.
  • Used as an internal standard in LC-MS/MS and GC-MS workflows.
  • Supplied as a small quantity solid to minimize waste for method development.
  • Intended for research use only; not for human or clinical applications.
Catalog No. 50-002-84820 Supplier Medchemexpress LLC Supplier No. HYW65434225MG
Add to Cart
Add to Cart
Spermine-d20 is a deuterium-labeled isotopologue of the polyamine spermine supplied as a research-grade stable isotope standard for analytical and biological studies. It is intended for use as an internal standard, tracer, or reference material in mass spectrometry and metabolomics. For research use only.
  • Deuterium-labeled standard for isotope dilution and tracer studies.
  • High purity suitable for analytical applications.
  • Used as an internal standard in LC-MS/MS and GC-MS workflows.
  • Supplied as a small quantity solid to minimize waste for method development.
  • Intended for research use only; not for human or clinical applications.