Learn More
Medchemexpress LLC Spermine-d20 (deuterium-labeled spermine) | 71-44-3 | >95.0% | 222.46 g/mol | C10H6D20N4 | 50 MG
SDP
Catalog No. 5000284311
Encompass_Preferred
Spermine-d20 is a deuterium-labeled form of spermine used as a stable isotope internal standard and tracer for analytical and biochemical workflows. It provides a predictable mass shift while retaining the chemical properties of unlabeled spermine, enabling accurate quantitation and tracing in LC-MS and isotopic studies.
  • Deuterium-labeled internal standard for mass spectrometry quantitation.
  • Suitable as a tracer in metabolic and pharmacokinetic studies.
  • Maintains native chemical behavior while providing mass differentiation.
  • Typical purity >95% (HPLC), suitable for analytical use.
  • Available in small pack sizes for laboratory use.
Catalog No. 50-002-84311 Supplier Medchemexpress LLC Supplier No. HYW65434250MG
Spermine-d20 is a deuterium-labeled form of spermine used as a stable isotope internal standard and tracer for analytical and biochemical workflows. It provides a predictable mass shift while retaining the chemical properties of unlabeled spermine, enabling accurate quantitation and tracing in LC-MS and isotopic studies.
  • Deuterium-labeled internal standard for mass spectrometry quantitation.
  • Suitable as a tracer in metabolic and pharmacokinetic studies.
  • Maintains native chemical behavior while providing mass differentiation.
  • Typical purity >95% (HPLC), suitable for analytical use.
  • Available in small pack sizes for laboratory use.