Learn More
Medchemexpress LLC Spermine-d8 tetrahydrochloride | 1173022-85-9 | 95% | 5 MG
SDP
Catalog No. 5000412862
Encompass_Preferred
Spermine-d8 (tetrahydrochloride) is the deuterium labeled Spermine tetrahydrochloride, a polyamine nitric oxide donor found in mammalian tissues, plants, bacteria, ribosomes, and bacteriophage. It acts as a natural antioxidant and anti-inflammatory agent, and is known to inhibit certain bacterial cultures. It can reversibly inhibit DNA synthetic response and mixed lymphocyte response.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for quantitation during drug development
  • Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-12862 Supplier Medchemexpress LLC Supplier No. HYB1777AS5MG
Spermine-d8 (tetrahydrochloride) is the deuterium labeled Spermine tetrahydrochloride, a polyamine nitric oxide donor found in mammalian tissues, plants, bacteria, ribosomes, and bacteriophage. It acts as a natural antioxidant and anti-inflammatory agent, and is known to inhibit certain bacterial cultures. It can reversibly inhibit DNA synthetic response and mixed lymphocyte response.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for quantitation during drug development
  • Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs