Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spermine dihydrate, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018522 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8522 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8522 Supplier MilliporeSigma Supelco Supplier No. 855885G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

MDL Number MFCD01459904
Synonym Gerontine; Musculamine; N,N′-Bis(3-aminopropyl)-1,4-diaminobutane; Neuridine

Specifications

Melting Point 63°C to 64°C
CAS 403982-64-9
pH 12.0 to 13.5 (25°C, 1 M aq. soln.)
Linear Formula NH2(CH2)3 NH(CH2)4 NH(CH2)3 NH2 · 2H2 O
Quantity 5 g
Solubility Information Water: 1 M (at 20°C, clear, colorless)
Formula Weight 238.37
Percent Purity ≥99.5% (GC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.