Learn More
Enzo Life Sciences Spermine NONOate (5mg). CAS: 136587-13-8
SDP
Catalog No. 502009136
Encompass_Preferred
Nitric oxide (NO) donor. A convenient reagent for preparing aqueous solutions of NO. Alternative name: SPER/NO, (Z)-1-N-[3-Aminopropyl]-N-[4-(3-aminopropylammonio)butyl]-amino-diazen-1-ium-1,2-diolate. Purity: ≥97% (NMR). Solubility: Soluble in water, methanol or 100% ethanol. Long Term Storage: -80°C.
Catalog No. 50-200-9136 Supplier Enzo Life Sciences Supplier No. ALX430013M005
Nitric oxide (NO) donor. A convenient reagent for preparing aqueous solutions of NO. Alternative name: SPER/NO, (Z)-1-N-[3-Aminopropyl]-N-[4-(3-aminopropylammonio)butyl]-amino-diazen-1-ium-1,2-diolate. Purity: ≥97% (NMR). Solubility: Soluble in water, methanol or 100% ethanol. Long Term Storage: -80°C.