Learn More
Abcam Spermine tetrahydrochloride, polyamine used for precipitation of DNA, 1G
Catalog No. 502704599
Encompass_Preferred
MW 348.19 g/mol. Spermine is a biogenic polyamine formed from spermidine and occurs in almost all tissues. It is essential for both neoplastic and normal tissue growth. Spermine is associated with nucleic acids, particularly in viruses, and is thought to stabilize the helical structure. Spermine tetrahydrochloride is used for precipitation of DNA from low salt aqueous buffers.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4599 Supplier Abcam Supplier No. AB1465231G
MW 348.19 g/mol. Spermine is a biogenic polyamine formed from spermidine and occurs in almost all tissues. It is essential for both neoplastic and normal tissue growth. Spermine is associated with nucleic acids, particularly in viruses, and is thought to stabilize the helical structure. Spermine tetrahydrochloride is used for precipitation of DNA from low salt aqueous buffers.
The product is subject to the following: Abcam Restricted Use Statement