Learn More
Medchemexpress LLC Spesolimab, 5mg
SDP
Supplier:  Medchemexpress LLC HYP993965MG
Encompass_Preferred
Spesolimab (BI 655130) is an anti-IL-36R antibody. IL-36 involves in immune system, as Spesolimab can be used for palmoplantar pustulosis (PPP) research. Spesolimab is associated with the reduction of biomarkers linked to innate, Th1/Th17, and neutrophilic pathways.
Catalog No. NC2810508