Learn More
Microsolv Technology Corporation Septa, 20mm, Headspace, Clear Silicone Rubber / White PTFE. For use in 20mm Crimp Caps, sold separately. 1000/CS.
Catalog No. 502270507
Encompass_Preferred
Silicone Rubber / PTFE Septa 60mil, 50mil rubber and 10mil PTFE. This Septa is good for higher temperature analyses. Temperature range from -60⁰ to 200⁰C. Use in Aluminum or Steel Crimp Caps for Headspace autosamplers. Supplied in Polyethylene Bags.
This item is not returnable. View return policy
Catalog No. 50-227-0507 Supplier Microsolv Technology Corporation Supplier No. 960201X5010
Silicone Rubber / PTFE Septa 60mil, 50mil rubber and 10mil PTFE. This Septa is good for higher temperature analyses. Temperature range from -60⁰ to 200⁰C. Use in Aluminum or Steel Crimp Caps for Headspace autosamplers. Supplied in Polyethylene Bags.