Learn More
Microsolv Technology Corporation Septa, 22mm, Translucent Silicone Rubber / Natural PTFE. Ultra Low Bleed. For use in 24mm Caps, sold separately. 1000/CS.
Catalog No. 502270510
Encompass_Preferred
Silicone Rubber / PTFE Septa 2.5mm thick. This Septa is good for storage or EPA Testing due to the high purity of the rubber and PTFE. Temperature range from -60⁰ to 200⁰C. Use in screw caps that have 24mm or ND2 threads. Supplied in a polyethylene Bags.
This item is not returnable. View return policy
Catalog No. 50-227-0510 Supplier Microsolv Technology Corporation Supplier No. 9704109
Silicone Rubber / PTFE Septa 2.5mm thick. This Septa is good for storage or EPA Testing due to the high purity of the rubber and PTFE. Temperature range from -60⁰ to 200⁰C. Use in screw caps that have 24mm or ND2 threads. Supplied in a polyethylene Bags.