Learn More
Description
Meeting all your QuEChERS needs with general purpose kits suitable for removal of polar organic acids, some low to moderate levels of sugars and lipids.
- US (AOAC Method 2007.01)
Specifications
Specifications
| Packaging | 100 pk. |
| Methods | AOAC 2007.01 |
| Analyte or Component Names | 50mg PSA, 150mg MgSO4 |
| Technique | Pesticide Extraction |
| Type | PSA Dispersive SPE Clean-up Tube |
| Element | PSA, MgSO4 |
| Quantity | 2 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
