Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPEX CertiPrep™ SPEXQuE™ GC/MS QC Standards
SDP

Catalog No. QUECRMSQC2 Shop All Spex CertiPrep Products
Change view
Click to view available options
Type:
GC/MS QC Standard 1
GC/MS QC Standard 2
Concentration:
100 μg/mL
50 μg/mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Concentration
QUECRMSQC2 GC/MS QC Standard 2 100 μg/mL
QUECRMSQC1 GC/MS QC Standard 1 50 μg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. QUECRMSQC2 Supplier SPEX CertiPrep™ Supplier No. QUECRMSQC2

missing translation for 'validMainframeMsg'

Sensible solutions for pesticide extraction

  • Solvent: Acetonitrile
  • Volume: 5mL

Specifications

Concentration 100 μg/mL
For Use With (Application) Sensible Solutions for Pesticide Extraction
Type GC/MS QC Standard 2
Volume (Metric) 5 mL
Material Anthracene
Solvents Acetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.