Learn More
Cole-Parmer™ Replacement End Plugs for Mid-Size Grinding Vial Set, Polycarbonate
Catalog No. 6885E
Change view
Click to view available options
Material:
Polycarbonate
Catalog No. Material
68-85E Polycarbonate
1 options
Catalog No. 68-85E Supplier Cole-Parmer™ Supplier No. 6885E
Add to Cart
Add to Cart
Replacement polycarbonate end plugs for (6885) Mid-Size Poly-Vial Set. Sold in pairs.
  • Ideal for grinding soft plant and animal tissues and eliminates metal contamination.
  • Withstands liquid nitrogen temperatures.
  • May be cleaned with hot water and detergent or RNase/DNase-free solution.
  • Made of durable polycarbonate.
Specifications
For Use With (Equipment) 6885 Mid-Size Grinding Vial Set (61043-13)
For Use With (Application) For Grinding Soft Plant and Animal Tissues Without Metal Contamination
Material Polycarbonate