Learn More
Cole-Parmer™ Grinding Vial Set, Mid-Size Vial, PC Cylinder and End Plugs, PC Encapsulated Steel Impactor
Description
- Grinds one sample at a time in the Large Freezer/Mill™ when used with the 6886 Mid-Size Grinding Vial Adapter.
- Sample grinding capacity 3 g – 25 g.
- Ideal for grinding soft plant and animal tissues and eliminates metal contamination.
- Withstands liquid nitrogen temperatures.
- Closed grinding vial prevents sample cross-contamination, retains volatiles, and controls hazardous or critical samples.
- Transparent plastic center cylinder allows visual checking of the grinding progress.
- Polycarbonate center cylinders are disposable.
- Polycarbonate end plugs, impactor, and center cylinders may be cleaned with hot water and detergent or RNase/DNase-free solution.
- Compatible with 6804 Large Extractor/Vial Opener when used in conjunction with 6884 Mid-Size Grinding Vial Adapter.
Specifications
Specifications
| Capacity (English) | 0.85 oz. |
| Capacity (Metric) | 25 mL |
| For Use With (Equipment) | 6870 Freezer/Mill |
| For Use With (Application) | For Grinding Soft Tissues Without Metal Contamination |
| Material | Polycarbonate |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.