Learn More
Cole-Parmer™ Replacement Impactor for Mid-Size Grinding Vial Set (61043-13), PC Encapsulated Steel
Catalog No. 6885P
Change view
Click to view available options
Material:
Polycarbonate
Catalog No. Material
68-85P Polycarbonate
1 options
Catalog No. 68-85P Supplier Cole-Parmer™ Supplier No. 6885P
Add to Cart
Add to Cart
Replacement polycarbonate encapsulated impactor for (6885) Mid-Size Poly-Vial Set.
  • Ideal for grinding soft plant and animal tissues and eliminates metal contamination.
  • Withstands liquid nitrogen temperatures.
  • May be cleaned with hot water and detergent or RNase/DNase-free solution.
  • Made of durable polycarbonate.
Specifications
Capacity (English) 0.8 oz.
Capacity (Metric) 25 mL
For Use With (Equipment) 6885 Mid-Size Grinding Vial Set (61043-13)
For Use With (Application) For Grinding Soft Plant and Animal Tissues Without Metal Contamination
Material Polycarbonate